Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate), Reagents

Catalog No. p-200021071
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg

Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate)
CAS 7558-79-4
Color Colorless/White
pH 8.4 to 9.6
Physical Form Solid
Quantity 100 g
Molecular Formula Na2HPO4
Synonym Disodium Hydrogen Phosphate
Molecular Weight (g/mol) 141.9588
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.066
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.