Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Phosphate Dibasic, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Description
- Suitable for Buffer Solutions
Specifications
Quantity | 500 g |
IUPAC Name | disodium;hydrogen phosphate |
Formula Weight | 141.96 |
Grade | Meets ACS reagent chemical requirements |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.07kg |
Chemical Name or Material | Sodium Phosphate Dibasic, Anhydrous |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction