Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, Anhydrous, BiotechGrade, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
7558-79-4
HNa2O4P
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O
7558-79-4
HNa2O4P
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O

S1401, 7558-79-4, Na2HPO4

Spectrum™ Sodium Phosphate, Dibasic, Anhydrous, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.

Grade Biotech
CAS Max % 1
Appearance White powder
Arsenic (As) 1.5e-005
Percent Purity 98.0 to 100.5%
DNase RNase Protease act None detected
Heavy Metals (as Pb) 1e-005
Insoluble Matter 0.0038
Iron (Fe) 2e-005
Loss on Drying 0.05
Odor Odorless
Packaging Poly Bottle
Sulfate 0.0019
Quantity 500 g
Show More Show Less