Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, Anhydrous, Powder, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7558-79-4
HNa2O4P
141.957
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
CHEBI:34683

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications
CAS 7558-79-4
Molecular Formula HNa2O4P
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;hydrogen phosphate
Molecular Weight (g/mol) 141.957
PubChem CID 24203
ChEBI CHEBI:34683
Formula Weight 141.96
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 2.07kg
Quantity 500g
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous
Grade Ultrapure Bioreagent
Show More Show Less