Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, Anhydrous, U.S.P. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500g
1kg
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7558-79-4
HNa2O4P
141.957
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
CHEBI:34683
disodium;hydrogen phosphate
OP(=O)([O-])[O-].[Na+].[Na+]
Formula Weight 141.96
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.07kg
Quantity 500g
Chemical Name or Material Sodium Phosphate, Dibasic, Anhydrous
Grade FCC/USP