Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate dibasic dihydrate, 98+%, Acros Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
10028-24-7
HNa2O42H2O
177.99
KDQPSPMLNJTZAL-UHFFFAOYSA-L
disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o
6451167
disodium;hydrogen phosphate;dihydrate
O.O.OP(=O)([O-])[O-].[Na+].[Na+]
10028-24-7
HNa2O42H2O
177.99
KDQPSPMLNJTZAL-UHFFFAOYSA-L
disodium hydrogen phosphate dihydrate,disodium phosphate dihydrate,sodium phosphate dibasic dihydrate,di-sodium hydrogen phosphate dihydrate,phosphoric acid, disodium salt, dihydrate,sodium hydrogenphosphate dihydrate,sodium phosphate, dibasic, dihydrate,disodium hydrogenphosphate dihydrate,dinatrii phosphas dihydricus,2na.hpo4.2h2o
6451167
disodium;hydrogen phosphate;dihydrate
O.O.OP(=O)([O-])[O-].[Na+].[Na+]
Assay Percent Range 98+%
Linear Formula Na2HPO4·2H2O
Merck Index 15,8789
Solubility Information Solubility in water: 93g/L(20°C)
Formula Weight 177.99
Percent Purity 98+%
Grade Analysis
Packaging Glass bottle
Solubility Solubility in water: 93g/L(20°C).
Melting Point 243.0°C to 245.0°C
Quantity 1kg
Chemical Name or Material Sodium phosphate dibasic dihydrate
Show More Show Less