Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, monobasic, 99%, for analysis, anhydrous, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
10kg
Packaging:
Plastic bottle
Plastic drum
7558-80-7
H2NaPO4
119.98
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate
23672064
CHEBI:37585
sodium;dihydrogen phosphate
OP(=O)(O)[O-].[Na+]
7558-80-7
H2NaPO4
119.98
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
monosodium phosphate,sodium dihydrogen phosphate,sodium dihydrogenorthophosphate,sodium phosphate monobasic,sodium acid phosphate,sodium phosphate, monobasic,monosodium dihydrogen orthophosphate,sodium phosphate monobasic anhydrous,monosodium orthophosphate,monobasic sodium phosphate
23672064
CHEBI:37585
sodium;dihydrogen phosphate
OP(=O)(O)[O-].[Na+]
Biological buffer
Assay Percent Range 99%
Linear Formula NaH2PO4
Merck Index 15,879
Arsenic (As) 1ppm max.
Solubility Information Solubility in water: 850g/L (20°C).
Formula Weight 119.98
Physical Form Crystalline Powder
Percent Purity 99%
Grade Analysis
Calcium (Ca) 100ppm max.
Chloride 0.002% max
Copper (Cu) 10ppm max.
Insoluble in Water 0.05% max.
Insoluble Matter 0.05% max. (in water)
Iron (Fe) 10ppm max.
Lead (Pb) 10ppm max.
Magnesium (Mg) 100ppm max.
Nickel (Ni) 10ppm max.
Nitrogen Compounds 0.002% max.
Packaging Plastic bottle
Solubility Solubility in water: 850g/L (20°C).
Sulfate 0.005% max
Color White
pH 4.1 to 4.7 (5% soln.)
Quantity 250g
Chemical Name or Material Sodium phosphate, monobasic
Show More Show Less

EINECSNumber : 231-449-2