Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate, monobasic dihydrate, 98%, extra pure, Acros Organics

Click to view available options
Quantity:
1kg
5kg
25kg
13472-35-0
H2NaO42H2O
156.01
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium dihydrogen phosphate dihydrate,sodium phosphate monobasic dihydrate,monosodium phosphate dihydrate,sodium phosphate, monobasic, dihydrate,phosphoric acid, monosodium salt, dihydrate,sodium dihydrogenphosphate dihydrate,sodium phosphate nah2po4 dihydrate,monosodium dihydrogen phosphate dihydrate,sodium dihydrate dihydrogen phosphate,acmc-20ajoh
23673460
sodium;dihydrogen phosphate;dihydrate
O.O.OP(=O)(O)[O-].[Na+]
13472-35-0
H2NaO42H2O
156.01
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium dihydrogen phosphate dihydrate,sodium phosphate monobasic dihydrate,monosodium phosphate dihydrate,sodium phosphate, monobasic, dihydrate,phosphoric acid, monosodium salt, dihydrate,sodium dihydrogenphosphate dihydrate,sodium phosphate nah2po4 dihydrate,monosodium dihydrogen phosphate dihydrate,sodium dihydrate dihydrogen phosphate,acmc-20ajoh
23673460
sodium;dihydrogen phosphate;dihydrate
O.O.OP(=O)(O)[O-].[Na+]
Assay Percent Range 98%
Linear Formula NaH2PO4·2H2O
Merck Index 15,879
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 156.01
Percent Purity 98%
Grade Extra Pure
Packaging Fibre drum
Solubility Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Melting Point 60.0°C
Quantity 25kg
Chemical Name or Material Sodium phosphate, monobasic dihydrate
Show More Show Less