Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Crystal, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10049-21-5
H4NaO5P
137.991
BBMHARZCALWXSL-UHFFFAOYSA-M
516949

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications
SMILES O.OP(=O)(O)[O-].[Na+]
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Quantity 500g
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate
Grade Ultrapure Bioreagent