Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade, 98-103%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10049-21-5
H5NaO5P
139.00
AXCRQNJHVGYJII-UHFFFAOYSA-N
phosphoric acid hydrate sodium
O.[Na].OP(O)(O)=O
10049-21-5
H5NaO5P
139.00
AXCRQNJHVGYJII-UHFFFAOYSA-N
phosphoric acid hydrate sodium
O.[Na].OP(O)(O)=O

S1396, 10049-21-5, NaH2PO4•H2O

Spectrum™ Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.

CAS Max % 1
Appearance White granules
Arsenic (As) 3e-006
Percent Purity 98 to 103%
Fluoride 5e-005
Heavy Metals (as Pb) 1e-005
Insoluble Matter 0.002
Odor Odorless
Packaging Poly Bottle
Sulfate 0.0015
Water 10-15%
pH 4.1 to 4.5
Quantity 4 x 2.5 kg
Show More Show Less