Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Monobasic, Monohydrate, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
10049-21-5
H4NaO5P
137.991
BBMHARZCALWXSL-UHFFFAOYSA-M
sodium dihydrogen phosphate monohydrate,sodium phosphate monobasic monohydrate,monosodium phosphate monohydrate,unii-593yog76rn,phosphoric acid, monosodium salt, monohydrate,sodium phosphate monobasic hydrate,sodium dihydrogen phosphate hydrate,sodium dihydrogenphosphate monohydrate,sodium hydrate dihydrogen phosphate,pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.OP(=O)(O)[O-].[Na+]
10049-21-5
H4NaO5P
137.991
BBMHARZCALWXSL-UHFFFAOYSA-M
sodium dihydrogen phosphate monohydrate,sodium phosphate monobasic monohydrate,monosodium phosphate monohydrate,unii-593yog76rn,phosphoric acid, monosodium salt, monohydrate,sodium phosphate monobasic hydrate,sodium dihydrogen phosphate hydrate,sodium dihydrogenphosphate monohydrate,sodium hydrate dihydrogen phosphate,pubchem12707
516949
sodium;dihydrogen phosphate;hydrate
O.OP(=O)(O)[O-].[Na+]
Formula Weight 137.99
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 2.04kg
Quantity 2.5kg
Chemical Name or Material Sodium Phosphate, Monobasic, Monohydrate