Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium phosphonoformate hexahydrate, 98+%

Click to view available options
Quantity:
1g
5g
25g
34156-56-4
CH12Na3O11P
300.039
MFCD00150176
ILRVASBWNRYBFD-UHFFFAOYSA-K
foscarnet sodium hexahydrate,sodium phosphonatoformate hexahydrate,unii-964ys0oog1,foscarnet trisodium hexahydrate,phosphonoformic acid trisodium salt hexahydrate,trisodium phosphonoformte hexahydrate,sodium phosphonoformate tribasic hexahydrate,phosphonoformic acid, trisodium salt, hexahydrate,trisodium foscarnet 3-hexahydrate,formic acid, phosphono-, trisodium salt, hexahydrate
169569
CHEBI:60269
trisodium;phosphonatoformate;hexahydrate
C(=O)([O-])P(=O)([O-])[O-].O.O.O.O.O.O.[Na+].[Na+].[Na+]
34156-56-4
CH12Na3O11P
300.039
MFCD00150176
ILRVASBWNRYBFD-UHFFFAOYSA-K
foscarnet sodium hexahydrate,sodium phosphonatoformate hexahydrate,unii-964ys0oog1,foscarnet trisodium hexahydrate,phosphonoformic acid trisodium salt hexahydrate,trisodium phosphonoformte hexahydrate,sodium phosphonoformate tribasic hexahydrate,phosphonoformic acid, trisodium salt, hexahydrate,trisodium foscarnet 3-hexahydrate,formic acid, phosphono-, trisodium salt, hexahydrate
169569
CHEBI:60269
trisodium;phosphonatoformate;hexahydrate
C(=O)([O-])P(=O)([O-])[O-].O.O.O.O.O.O.[Na+].[Na+].[Na+]
Applications
Sodium phosphonoformate hexahydrate acts as an antiviral agent and reverse transcriptase inhibitor. It is also used to inhibit viral DNA polymerase. Further, it is used as a type II Pi transporter inhibitor.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
Quantity 1g
Solubility Information Soluble in water.
Formula Weight 300.08 (191.98 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Sodium phosphonoformate hexahydrate

Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)

RTECSNumber : SY8200000

TSCA : No

Recommended Storage : Ambient temperatures

RMF - Research Use or Further Mfg