Sodium pieces, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4306818

Quantity 50g

 View more versions of this product

Catalog No. AA4306818

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7440-23-5
    Physical Form Pieces, 3-12mm (0.1-0.5 in.)
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00085307
    Chemical Name or Material Sodium

    Specifications

    Quantity 50g
    Color Silver-gray
    DOT Information DOT Class: 4.3, Packaging Group: I
    EINECS Number 231-132-9
    Physical Form Pieces, 3-12mm (0.1-0.5 in.)
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00085307
    CAS 7440-23-5
    Odor Nearly odorless
    Health Hazard 1 H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.
    Appearance Silver Gray pieces
    Chemical Name or Material Sodium

    H260-H314-H318: In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. Causes serious eye damage.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit