Sodium, plasma standard solution, Na 10,000µg/mL, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 14400AE

Quantity 100mL

 View more versions of this product

Catalog No. AA14400AE

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula Na2CO3 in 5% HNO3
    MDL Number MFCD00085307
    Chemical Name or Material Sodium, plasma standard solution

    Specifications

    Quantity 100mL
    Color Clear
    DOT Information DOT Class: 8, Packaging Group: III
    Health Hazard 2 Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    Physical Form Liquid
    Appearance Clear liquid
    Molecular Formula Na2CO3 in 5% HNO3
    MDL Number MFCD00085307
    Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Sodium carbonate: 2.3%; Water: 92.7%
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Sodium, plasma standard solution

    Keep container in a well ventilated place. Keep away from combustible material. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit