Sodium, plasma standard solution, Na 1000µg/mL, Specpure™, Alfa Aesar™

Manufacturer: Alfa Aesar 13832AD

Quantity 50mL

 View more versions of this product

Catalog No. AA13832AD

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula Na2CO3 in 5% HNO3
    MDL Number MFCD00085307
    Chemical Name or Material Na 1000μg/mL

    Specifications

    Quantity 50mL
    Color Clear
    Physical Form Liquid
    MDL Number MFCD00085307
    Vapor Pressure 23hPa (17mm Hg) at 20°C
    Molecular Formula Na2CO3 in 5% HNO3
    Chemical Name or Material Na 1000μg/mL

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit