Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Polyphosphates, Glassy, FCC, 60-71%, Spectrum™

Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
68915-31-1
H2Na2O7P2
221.94
GYQBBRRVRKFJRG-UHFFFAOYSA-L
disodium hydrogen (hydrogen phosphonatooxy)phosphonate
[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O
68915-31-1
H2Na2O7P2
221.94
GYQBBRRVRKFJRG-UHFFFAOYSA-L
disodium hydrogen (hydrogen phosphonatooxy)phosphonate
[Na+].[Na+].OP([O-])(=O)OP(O)([O-])=O

SO169, 68915-31-1, Approx. (NaPO3)n

Spectrum™ Sodium Polyphosphates, Glassy, FCC is used as an acidity regulator, emulsifier and buffering agent, and as a sequestering agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 60 to 71%
Grade FCC
Fluoride 5e-005
Insoluble Matter 0.001
Lead (Pb) 4 mg/kg
Packaging Fiber Drum
Quantity 45 kg