Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Polystyrene Sulfonate, USP, Spectrum™

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

S1408, 9003-59-2

Spectrum™ Sodium Polystyrene Sulfonate, USP is used to treat hyperkalemia (increased amounts of potassium in the body). All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Water 0.1
Quantity 5 kg
CAS 9003-59-2
CAS Max % 1
Grade USP