Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Potassium ATPase Alpha 2 Antibody (SR1026), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Sodium Potassium ATPase Alpha 2 |
---|---|
Clone | SR1026 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Sodium Potassium ATPase Alpha 2 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Sodium Potassium ATPase Alpha 2 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
alpha-A(+) catalytic polypeptide, ATPase, Na+/K+ transporting, alpha 2 (+) polypeptide, ATPase, Na+/K+ transporting, alpha 2 polypeptide, EC 3.6.3, EC 3.6.3.9, KIAA0778, MGC59864, MHP2, migraine, hemiplegic 2, Na(+)/K(+) ATPase alpha-2 subunit, Na+/K+ ATPase, alpha-B polypeptide, Sodium pump subunit alpha-2, sodium/potassium-transporting ATPase alpha-2 chain, sodium/potassium-transporting ATPase subunit alpha-2, sodium-potassium ATPase | |
A synthesized peptide derived from human Sodium Potassium ATPase Alpha 2 (Uniprot #: P50993) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1026 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
477 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title