Alfa Aesar™ Sodium potassium (K2Na)-silica gel, 35-40% alkali metal in silica gel, Stage I

Manufacturer: Alfa Aesar™ 4489022

Quantity 100g

 View more versions of this product

Catalog No. AA4489022

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD08705348
    UN Number UN2813
    Chemical Name or Material Sodium potassium (K2Na)-silica gel, Stage I

    Specifications

    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.7g/mL
    Physical Form Powder
    UN Number UN2813
    Health Hazard 1 Danger
    Sensitivity Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
    MDL Number MFCD08705348
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Color Black
    Concentration 35 to 40% alkali metal in silica gel
    Quantity 100g
    Chemical Name or Material Sodium potassium (K2Na)-silica gel, Stage I

    Description

    • Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit