Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Sodium potassium (K2Na)-silica gel, 35-40% alkali metal in silica gel, Stage I

Manufacturer:  Alfa Aesar™ 4489006

Quantity 5g

 View more versions of this product

Catalog No. AA4489006


  Check Availability
Add to cart

Specifications

Description

  • Reductively produce radical anions and dianions, dechlorinate alkyl and aryl halides and desulfurize compounds
Description & Specifications

Chemical Identifiers

MFCD08705348
UN2813
Sodium potassium (K2Na)-silica gel, Stage I

Specifications

0.7g/mL
MFCD08705348
Black
5g
Sodium potassium (K2Na)-silica gel, Stage I
Powder
UN2813
35 to 40% alkali metal in silica gel
Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261-H314-H318
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Handle under inert gas.
Protect from moisture.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit