Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Propionate, NF, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
6700-17-0
C3H7NaO3
114.08
HOAUAOBUGFYWMK-UHFFFAOYSA-M
sodium hydrate propanoate
O.[Na+].CCC([O-])=O
6700-17-0
C3H7NaO3
114.08
HOAUAOBUGFYWMK-UHFFFAOYSA-M
sodium hydrate propanoate
O.[Na+].CCC([O-])=O

S1418, 6700-17-0, C3H5NaO2•xH2O

Spectrum™ Sodium Propionate, NF is used as an antiseptic agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Quantity 12 kg
Water 0.01
CAS Max % 1
Percent Purity 99 to 100.5%
Grade NF