Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
13472-36-1
Na4O7P2·10H2O
446.05
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate,unii-iy3dkb96qw,tetrasodium pyrophosphate decahydrate,diphosphoric acid, tetrasodium salt, decahydrate,iy3dkb96qw,pyrophosphate sodium salt decahydrate,pyrophosphate inorganic,colostrum,sodium phosphate pyro,sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
13472-36-1
Na4O7P2·10H2O
446.05
VZWGHDYJGOMEKT-UHFFFAOYSA-J
sodium pyrophosphate decahydrate,unii-iy3dkb96qw,tetrasodium pyrophosphate decahydrate,diphosphoric acid, tetrasodium salt, decahydrate,iy3dkb96qw,pyrophosphate sodium salt decahydrate,pyrophosphate inorganic,colostrum,sodium phosphate pyro,sodium pyrophosphate na4p2o7 decahydrate
3084150
tetrasodium;phosphonato phosphate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]
Assay Percent Range 99-103%
Linear Formula Na4P2O7·10H2O
Solubility Information Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia,insoluble in alcohol
Formula Weight 446.05
Percent Purity 99-103%
Grade ACS Reagent
Chloride 0.002% max
Insoluble Matter 0.01% max
Packaging Plastic bottle
Solubility Solubility in water: 62g/L (20°C). Other solubilities: insoluble in ammonia, insoluble in alcohol
Sulfate 0.005% max
Boiling Point 93.8°C
Melting Point 80.0°C
Quantity 2.5kg
Chemical Name or Material Sodium pyrophosphate decahydrate
Show More Show Less

GHS Signal Word: Warning