Learn More
Sodium selenate, anhydrous, 99.8+% (metals basis), Thermo Scientific Chemicals
Description
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 32° (transition) |
Color | Purple |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99.8+% (metals basis) |
Solubility Information | Soluble in water. |
Formula Weight | 188.94 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | White powder |
Show More |
Safety and Handling
Hazard Category | H300-H331-H373 |
Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
DOTInformation | DOT Class: 6.1, Packaging Group: I |
EINECSNumber | 236-501-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.