Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenate, Anhydrous, Crystal, 97%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
13410-01-0
Na2O4Se
188.95
MHQOTKLEMKRJIR-UHFFFAOYSA-L
disodium selenate
[Na+].[Na+].[O-][Se]([O-])(=O)=O
13410-01-0
Na2O4Se
188.95
MHQOTKLEMKRJIR-UHFFFAOYSA-L
disodium selenate
[Na+].[Na+].[O-][Se]([O-])(=O)=O

S1428, 13410-01-0, Na2SeO4

Spectrum™ Sodium Selenate, Anhydrous, Crystal has several uses, including the manufacturing of glass, specifically red-hued glass, but can also produce clear glass when mixed with ferrous oxides. It's a white or gray solid substance that is highly toxic. Regardless of the toxicity, sodium selenate is used in vitamin supplements as a trace mineral additive and is being investigated as a potential anti-Alzheimers drug candidate.

Grade Reagent
CAS Max % 1
Percent Purity 97%
Packaging Poly Pail
Quantity 5 kg