Learn More
Sodium selenate decahydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 35°C (decomposition) |
Physical Form | Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.9% (metals basis) |
UN Number | UN2630 |
Merck Index | 14,8671 |
Solubility Information | Soluble in water. |
Formula Weight | 369.09 (188.94 Anhydrous) |
Percent Purity | 99.9% |
Density | 1.600 g/mL |
Show More |
Safety and Handling
Hazard Category | H301+H331-H373 |
Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.