Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium selenate decahydrate, 99.9% (metals basis)

Manufacturer:  Alfa Aesar 1423914

Quantity 25g

 View more versions of this product

Catalog No. AA1423914


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10102-23-5
369.09 (188.94 Anhydrous)
UN2630
Sodium selenate decahydrate
Na2SeO4·10H2O
MFCD00149174
14,8671

Specifications

10102-23-5
99.9% (metals basis)
369.09 (188.94 Anhydrous)
UN2630
Soluble in water
25g
99.9%, (metals basis)
Crystalline
Na2SeO4·10H2O
MFCD00149174
14,8671
1.6g/mL
Sodium selenate decahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H331-H373

GHS P Statement
P260-P261-P264-P301+P310-P304+P340-P311-P321-P405-P403+P233-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit