Alfa Aesar™ Sodium selenite, anhydrous, 99% min, typically 99.75% min (metals basis)

In glass manufacturing

Manufacturer: Alfa Aesar™ 1258536

Quantity 500g

 View more versions of this product

Catalog No. AA1258536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-18-8
    Linear Formula Na2SeO3
    Formula Weight 172.94
    MDL Number MFCD00003489
    UN Number UN2630
    Merck Index 14,8673
    Synonym Selenious acid disodium salt
    Chemical Name or Material Sodium selenite, Anhydrous

    Specifications

    Formula Weight 172.94
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Solubility Information Freely soluble in water; Insoluble in alcohol
    Health Hazard 2 GHS H Statement
    H300-H331-H317
    Fatal if swallowed.
    Toxic if inhaled.
    May cause an allergic skin reaction.
    RTECS Number VS7350000
    TSCA TSCA
    MDL Number MFCD00003489
    UN Number UN2630
    Synonym Selenious acid disodium salt
    Melting Point >350°C
    pH 9
    Health Hazard 1 Danger
    Linear Formula Na2SeO3
    CAS 10102-18-8
    Merck Index 14,8673
    Physical Form Powder
    Sensitivity Hygroscopic
    Color White
    Quantity 500g
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
    Chemical Name or Material Sodium selenite, Anhydrous
    Assay ≤99%, Typically ≤99.75% (metals basis)

    GHS H Statement
    H300-H331-H317
    Fatal if swallowed.
    Toxic if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P310-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit