Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sesquicarbonate, Dihydrate, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
533-96-0
C2HNa3O6
189.99
WCTAGTRAWPDFQO-UHFFFAOYSA-K
trisodium hydrogen carbonate carbonate
[Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O
533-96-0
C2HNa3O6
189.99
WCTAGTRAWPDFQO-UHFFFAOYSA-K
trisodium hydrogen carbonate carbonate
[Na+].[Na+].[Na+].OC([O-])=O.[O-]C([O-])=O

S1357, 533-96-0, Na2CO3•NaHCO3•2H2O

Spectrum™ Sodium Sesquicarbonate, Dihydrate , with a systematic name trisodium hydrogen dicarbonate, is used as an alkalinity source for water treatment. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Grade Reagent
CAS Max % 1
Packaging Fiber Drum
Quantity 45 kg