Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Alfa Aesar™

Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds

Manufacturer: Alfa Aesar 4489322

Quantity 100g

 View more versions of this product

Catalog No. AA4489322

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Powder
    MDL Number MFCD01941718
    Chemical Name or Material Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I

    Specifications

    Physical Form Powder
    Quantity 100g
    Color Black
    DOT Information DOT Class: 4.3, Packing Group: II
    Sensitivity Moisture sensitive
    MDL Number MFCD01941718
    Health Hazard 1 H228-H261-H314-H318: Flammable solid. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Causes serious eye damage.
    Health Hazard 2 Keep container tightly closed and dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing
    Chemical Name or Material Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I

    Keep container tightly closed and dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Take precautionary measures against static discharges. Wear suitable protective clothing

    H228-H261-H314-H318: Flammable solid. In contact with water releases flammable gas. Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit