Sodium-silica gel, 35-40% alkali metal in silica gel, Stage I, Alfa Aesar™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds
Manufacturer: Alfa Aesar 4489314
Quantity 25g
 View more versions of this product
Catalog No. AA4489314
  Check Availability
Add to Cart
Related Products
Product Content Feedback