Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II, Alfa Aesar™

Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds. Also Ideal For Reduction of Water To Produce Hydrogen Gas, Solvent Drying and Anti-Oxidant

Manufacturer:  Alfa Aesar 4489414

Quantity 25g

 View more versions of this product

Catalog No. AA4489414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Powder
MFCD01941718
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II

Specifications

Powder
0.7g/cm3
Black
Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II
MFCD01941718
Moisture sensitive
25g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228-H261-H314-H318

GHS P Statement
P210-P231+P232-P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P405-P501a

Danger

DOTInformation : DOT Class: 4.3, Packing Group: II

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit