Sodium-silica gel, 35-40% alkali metal in silica gel, Stage II, Alfa Aesar™
Reductively Produces Radical Anions and Dianions, Dechlorinates Alkyl and Aryl Halides and Desulfurize Compounds. Also Ideal For Reduction of Water To Produce Hydrogen Gas, Solvent Drying and Anti-Oxidant
Manufacturer: Alfa Aesar 4489406
Quantity 5g
 View more versions of this product
Catalog No. AA4489406
  Check Availability
Add to Cart
Related Products
Product Content Feedback