Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Silicate, 42° Be (Water Glass), Reagents

Catalog No. 01336968
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
HDPE Winpak
Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-968 200 L Poly Drum
01-336-970 10 L Cubitainer
01-336-969 1 L Poly Bottle
01-336-967 20 L HDPE Winpak
01-336-966 20 L Cubitainer
01-336-965 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-336-968 Supplier Reagents Holdings Llc Supplier No. C2325000200E
Only null left
Add to Cart
Add to Cart

Sodium Silicate, 42° Be’ (Water Glass), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1344-09-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Water Glass
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Melting Point -7°C
Color Colorless/White
Physical Form Liquid
Quantity 200 L
Grade Reagent
Packaging Poly Drum
Specific Gravity 1.408
Chemical Name or Material Sodium Silicate, 42° Be’ (Water Glass)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.