Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Silicate, Technical Grade, 40 to 42°Baumé, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
| Color | Colorless or White |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solubility Information | Solubility in Water: Infinite |
| Molecular Weight (g/mol) | 212.14 |
| Grade | Technical |
| Odor | Odorless |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Silicate, 40 to 42°Be |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction