Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stannate, Granular, Reagent, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
12027-70-2
H6Na2O6Sn
266.73
PMPBLIIMRRPPEO-UHFFFAOYSA-H
tin(4+) disodium hexahydroxide
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4]
12027-70-2
H6Na2O6Sn
266.73
PMPBLIIMRRPPEO-UHFFFAOYSA-H
tin(4+) disodium hexahydroxide
[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Na+].[Na+].[Sn+4]

S1445, 12027-70-2, Na2SnO3•3H2O

Spectrum™ Sodium Stannate, Granular, Reagent is a colorless salt that occurs when tin is dissolved in sodium hydroxide. It is an inorganic compound that is used as a hydrogen peroxide stabilizer. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Grade Reagent
CAS Max % 1
Packaging Poly Pail
Tin (Sn) 0.415
Quantity 12 kg