Sodium stearate

Manufacturer: Alfa Aesar 39648A1

Quantity 1kg

 View more versions of this product

Catalog No. AA39648A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 822-16-2
    Molecular Formula NaCO2(CH2)16CH3
    Formula Weight 306.46
    MDL Number MFCD00036404
    Merck Index 14,8678
    Synonym Sodium octadecanoate
    Chemical Name or Material Sodium stearate

    Specifications

    Formula Weight 306.46
    Quantity 1kg
    TSCA TSCA
    Merck Index 14,8678
    Solubility Information Slowly soluble in cold water; Solubility increases with temperature
    Physical Form Powder
    Molecular Formula NaCO2(CH2)16CH3
    MDL Number MFCD00036404
    CAS 822-16-2
    Synonym Sodium octadecanoate
    Chemical Name or Material Sodium stearate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit