Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stearate, Powder, NF, 90%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
822-16-2
C18H35NaO2
306.47
RYYKJJJTJZKILX-UHFFFAOYSA-M
sodium octadecanoate
[Na+].CCCCCCCCCCCCCCCCCC([O-])=O
822-16-2
C18H35NaO2
306.47
RYYKJJJTJZKILX-UHFFFAOYSA-M
sodium octadecanoate
[Na+].CCCCCCCCCCCCCCCCCC([O-])=O

S1447, 822-16-2, C18H35NaO2

Spectrum™ Sodium Stearate, Powder, NF is used in the pharmaceutical industry as a surfactant to aid the solubility of hydrophobic compounds. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade NF
CAS Max % 1
Percent Purity 90%
Loss on Drying 0.05
Packaging Fiber Drum
Quantity 12 kg