Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stearyl Fumarate, NF, 99-101.5%, Spectrum™

Click to view available options
Quantity:
500 g
25 kg
Packaging:
Fiber Drum
Poly Pail
4070-80-8
C22H39NaO4
390.54
STFSJTPVIIDAQX-LTRPLHCISA-M
sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate
[Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O
4070-80-8
C22H39NaO4
390.54
STFSJTPVIIDAQX-LTRPLHCISA-M
sodium (2E)-4-(octadecyloxy)-4-oxobut-2-enoate
[Na+].CCCCCCCCCCCCCCCCCCOC(=O)\C=C\C([O-])=O

S1448, 4070-80-8, C22H39NaO4

Spectrum™ Sodium Stearyl Fumarate, NF is a water-soluble tablet lubicant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Saponification Value 142.2-146.0
Quantity 25 kg
Water 0.05
CAS Max % 1
Percent Purity 99 to 101.5%
Grade NF