Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Succinate, Anhydrous, Reagent, 99-102%, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
Description
Spectrum™ Chemical Sodium Succinate, Anhydrous, Reagent is the sodium salt of a dicarboxylic acid which is used primarily as an acidity regulator and an industrial biotechnology raw material. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Specifications
CAS Max % | 1 |
Loss on Drying | 0.01 |
Packaging | Poly Pail |
Quantity | 2.5 kg |
Percent Purity | 99 to 102% |
Grade | Reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction