Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Succinate, Hexahydrate, Reagent, 97-102%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6106-21-4
C4H16Na2O10
270.14
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium butanedioate hexahydrate
O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O
6106-21-4
C4H16Na2O10
270.14
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium butanedioate hexahydrate
O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)CCC([O-])=O

S1449, 6106-21-4, C4H4Na2O4•6H2O

Spectrum™ Sodium Succinate, Hexahydrate, Reagent , also known as succinic acid, is a diprotic and dicarboxylic acid. It is involved in an energy-yielding process known as the citric acid cycle. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Grade Reagent
CAS Max % 1
Percent Purity 97 to 102%
Packaging Poly Pail
Quantity 2.5 kg