Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate, 99+%, for HPLC, anhydrous, Acros Organics

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15,8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide,insoluble in alcohol
Formula Weight 142.04
Percent Purity 99+%
Grade HPLC
Packaging Plastic bottle
Solubility Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
Boiling Point >1700.0°C
Melting Point 884.0°C
Quantity 50g
Chemical Name or Material Sodium sulfate
Show More Show Less

EINECSNumber : 231-820-9