Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate, ACS, 99.0% min. , Alfa Aesar™

Click to view available options
Quantity:
500g
2kg
10kg
7757-82-6
Na2O4S
142.036
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]
7757-82-6
Na2O4S
142.036
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]

For Use in Drying organic Liquids, in Kjeldahl Nitrogen Determination, in Manufacturing of Glass

Merck Index 14,8680
Solubility Information Soluble in water; Insoluble in alcoholSoluble in water,glycerol and hydrogen iodide; Insoluble in ethanol and many organic solvents.
Formula Weight 142.04
Physical Form Powder
Percent Purity ≥99.0%
Grade ACS
Density 2.680
Odor Odorless
Refractive Index 1.484
Sensitivity Hygroscopic
Melting Point 884°C
Quantity 500g
Chemical Name or Material Sodium sulfate
Show More Show Less

EINECSNumber : 231-820-9

RTECSNumber : WE1650000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen