Learn More
Sodium sulfate, ACS, 99.0% min, Thermo Scientific Chemicals
Description
In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 884°C |
Physical Form | Powder |
Quantity | 500 g |
Merck Index | 14,8680 |
Solubility Information | Soluble in water; Insoluble in alcoholSoluble in water,glycerol and hydrogen iodide; Insoluble in ethanol and many organic solvents. |
Formula Weight | 142.04 |
Percent Purity | ≥99.0% |
Grade | ACS |
Odor | Odorless |
Refractive Index | 1.484 |
Show More |
Safety and Handling
EINECSNumber | 231-820-9 |
RTECSNumber | WE1650000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.