Alfa Aesar™ Sodium sulfate, anhydrous, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 8761114

Quantity 25g

 View more versions of this product

Catalog No. AA8761114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7757-82-6
    Molecular Formula Na2O4S
    Linear Formula Na2SO4
    Formula Weight 142.04
    MDL Number MFCD00003504
    Merck Index 14,8680
    Chemical Name or Material Sodium sulfate, Anhydrous

    Specifications

    Physical Form Powder/Chunks
    Formula Weight 142.04
    Odor Odorless
    Molecular Formula Na2O4S
    CAS 7757-82-6
    Quantity 25g
    TSCA TSCA
    Refractive Index 1.4840
    Sensitivity Hygroscopic
    Linear Formula Na2SO4
    Merck Index 14,8680
    Color White
    MDL Number MFCD00003504
    Melting Point 884°C
    Density 2.680g/mL
    RTECS Number WE1650000
    Chemical Name or Material Sodium sulfate, Anhydrous
    Assay 99.99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit