Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, BiotechGrade, 99%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
Packaging:
Poly Bottle
Poly Pail
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O

S1457, 7757-82-6, Na2SO4

Spectrum™ Sodium Sulfate, Anhydrous, BiotechGrade is used in the laboratory as an inert drying agent for removing traces of water from organic solutions. It is only effective below 30°C, but is fairly inert and can be used with a variety of materials. Spectrum offers highly pure reagents suitable for biochemical research and analysis.

Grade Biotech
CAS Max % 1
Appearance White granules
Percent Purity 99%
Heavy Metals (as Pb) 5e-006
Insoluble Matter 0.0001
Iron (Fe) 1e-005
Odor Odorless
Packaging Poly Pail
Quantity 12 kg