Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, Granular Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
7757-82-6
Na2O4S
142.036
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]
7757-82-6
Na2O4S
142.036
PMZURENOXWZQFD-UHFFFAOYSA-L
sodium sulfate,disodium sulfate,salt cake,thenardite,sodium sulfate anhydrous,sodium sulphate,sodium sulfate, anhydrous,sulfuric acid disodium salt,disodium sulphate,sodium sulfate 2:1
24436
CHEBI:32149
disodium;sulfate
[O-]S(=O)(=O)[O-].[Na+].[Na+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 142.04
Physical Form Granular Powder
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.68kg
Quantity 500g
Chemical Name or Material Sodium Sulfate, Anhydrous