Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, Granular, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O
7757-82-6
Na2O4S
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
[Na+].[Na+].[O-]S([O-])(=O)=O

S1450, 7757-82-6, Na2SO4

Spectrum™ Sodium Sulfate, Anhydrous, Granular, Reagent, ACS is the sodium salt of sulfuric acid. It is a white, crystalline solid, and is used on a small scale in laboratories as an inert drying agent. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

CAS Max % 1
Percent Purity 99%
Calcium (Ca) 0.0001
Heavy Metals (as Pb) 5 ppm
Insoluble Matter 0.0001
Iron (Fe) 1e-005
Loss on Ignition 0.005
Magnesium (Mg) 5e-005
Packaging Amber Glass Bottle
Potassium (K) 0.0001
pH 5.2 to 9.2
Quantity 4 x 2.5 kg
Show More Show Less