Sodium Sulfate Anhydrous, min.99%, 12-60 Mesh, Granular, Ultra Resi-Analyzed, ACS Reagent, J.T.Baker™

Manufacturer: J.T.Baker 337507

Packaging Lined Fiber Drum
Quantity 12kg

 View more versions of this product

Catalog No. 14-650-503

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Granular
    Grade ACS Reagent Grade

    Specifications

    Formula Weight 142.04
    pH 5.2 to 9.2
    Packaging Lined Fiber Drum
    Specific Gravity 1L = 2.68kg
    Molecular Formula Na2SO4
    CAS 7757-82-6
    Quantity 12kg
    Physical Form Granular
    Heavy Metals (as Pb) max. 5ppm
    Insoluble Matter max. 0.01%
    Iron (Fe) max. 0.001%
    Loss on Ignition max. 0.5%
    Magnesium (Mg) max. 0.005%
    Nitrogen Compounds max. 5ppm
    Phosphate max. 0.001%
    pH Range 5.2 to 9.2
    Potassium (K) max. 0.01%
    Chloride max. 0.001%
    Chemical Name or Material Sodium Sulfate, Anhydrous
    Grade ACS Reagent Grade, Meets ACS reagent chemical requirements

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit