Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Decahydrate (Crystalline/Certified), Fisher Chemical

Catalog Number s419500
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
7727-73-3
H20Na2O14S
322.186
MFCD00149189
RSIJVJUOQBWMIM-UHFFFAOYSA-L
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
62649
CHEBI:32586
disodium;sulfate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
7727-73-3
H20Na2O14S
322.186
MFCD00149189
RSIJVJUOQBWMIM-UHFFFAOYSA-L
sodium sulfate decahydrate, glauber's salt, disodium sulfate decahydrate, sodium sulfate usp, sulfuric acid disodium salt, decahydrate, unii-0ypr65r21j, sodium sulfate na2so4 decahydrate, sodium sulphate decahydrate, sodium sulfate decahydrate na2so4.10h2o, disodium sulfate decahydrate na2so4.10h2o
62649
CHEBI:32586
disodium;sulfate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
Melting Point 884°C
Color White
pH 5.2 to 9.2
Physical Form Solid
Quantity 500 g
Formula Weight 322.19
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Density 1.46g/cm³
Chemical Name or Material Sodium Sulfate Decahydrate
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit