Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate decahydrate, extra pure, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
25kg
Packaging:
Fibre drum
Plastic bottle
7727-73-3
Na2O410H2O
322.2
MFCD00149189
RSIJVJUOQBWMIM-UHFFFAOYSA-L
sodium sulfate decahydrate,glauber's salt,disodium sulfate decahydrate,sodium sulfate usp,sulfuric acid disodium salt, decahydrate,unii-0ypr65r21j,sodium sulfate na2so4 decahydrate,sodium sulphate decahydrate,sodium sulfate decahydrate na2so4.10h2o,disodium sulfate decahydrate na2so4.10h2o
62649
CHEBI:32586
disodium;sulfate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
7727-73-3
Na2O410H2O
322.2
MFCD00149189
RSIJVJUOQBWMIM-UHFFFAOYSA-L
sodium sulfate decahydrate,glauber's salt,disodium sulfate decahydrate,sodium sulfate usp,sulfuric acid disodium salt, decahydrate,unii-0ypr65r21j,sodium sulfate na2so4 decahydrate,sodium sulphate decahydrate,sodium sulfate decahydrate na2so4.10h2o,disodium sulfate decahydrate na2so4.10h2o
62649
CHEBI:32586
disodium;sulfate;decahydrate
O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Na+].[Na+]
Linear Formula Na2SO4·10H2O
Fieser 01,1103
Merck Index 14,868
Solubility Information Solubility in water: soluble
Formula Weight 322.2
Physical Form Crystals or Crystalline Powder
Percent Purity ≥99.0%
Grade Extra Pure
Calcium (Ca) 0.01% max.
Chloride 0.02% max
Iron (Fe) 0.005% max.
Packaging Plastic bottle
Potassium (K) 0.01% max.
Solubility Solubility in water: soluble.
Color White
Melting Point 884.0°C
pH 5.2 to 9.2 (5% soln.)
Quantity 250g
Chemical Name or Material Sodium sulfate decahydrate
Show More Show Less